Skip to Content
Merck
All Photos(1)

Key Documents

8.14287

Sigma-Aldrich

5-tert-Butyl-m-xylene

for synthesis

Synonym(s):

5-tert-Butyl-m-xylene, 1-tert-Butyl-3,5-dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18
CAS Number:
Molecular Weight:
162.27
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-647-6

vapor pressure

0.4 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

560 °C (DIN 51794)

pH

7 (20 °C in H2O)

bp

207-209 °C/1013 hPa

mp

-22 °C

transition temp

flash point 84 °C (DIN 51758)

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C12H18/c1-9-6-10(2)8-11(7-9)12(3,4)5/h6-8H,1-5H3

InChI key

FZSPYHREEHYLCB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.864 - 0.866
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service