Skip to Content
Merck
All Photos(1)

Key Documents

8.14152

Sigma-Aldrich

trans-Stilbene

for synthesis

Synonym(s):

trans-Stilbene, trans-1,2-Diphenylethylene

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€147.00

€147.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€147.00

About This Item

Linear Formula:
C6H5CH=CHC6H5
CAS Number:
Molecular Weight:
180.25
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-098-5
NACRES:
NA.22

€147.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

crystals

bp

306-307 °C/1013 hPa

mp

121-124 °C

solubility

0.00029 g/L

density

1.14 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

c2(ccccc2)\C=C\c1ccccc1

InChI

1S/C14H12/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-12H/b12-11+

InChI key

PJANXHGTPQOBST-VAWYXSNFSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 122 °C
Melting range (upper value): ≤ 125 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service