Skip to Content
Merck
All Photos(1)

Key Documents

8.14126

Sigma-Aldrich

2-Mercaptobenzimidazole

for synthesis

Synonym(s):

2-Mercaptobenzimidazole, 2-Benzimidazolethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2S
CAS Number:
Molecular Weight:
150.20
MDL number:
UNSPSC Code:
12352105
EC Index Number:
209-502-6

Quality Level

Assay

≥98.0% (HPLC)

form

powder

autoignition temp.

480 °C

potency

300 mg/kg LD50, oral (Rat)

mp

285-290 °C

solubility

0.3 g/L

density

1.42 g/cm3

storage temp.

2-30°C

InChI

1S/C7H6N2S/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H2,8,9,10)

InChI key

YHMYGUUIMTVXNW-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Skin Sens. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

752.0 °F - closed cup

Flash Point(C)

> 400 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service