Skip to Content
Merck

Skip To

8.10595

Sodium 2-mercaptoethanesulfonate

for synthesis

Synonym(s):

Sodium 2-mercaptoethanesulfonate, 2-Mercaptoethanesulfonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Check Cart for Availability
€ 132,00
10 kg

About This Item

Empirical Formula (Hill Notation):
C2H5NaO3S2
CAS Number:
Molecular Weight:
164.18
UNSPSC Code:
12352107
EC Index Number:
243-285-9
NACRES:
NA.22
MDL number:
Form:
powder

€ 132,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

potency

4440 mg/kg LD50, oral (Rat)

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCS

InChI

1S/C2H6O3S2.Na/c3-7(4,5)2-1-6;/h6H,1-2H2,(H,3,4,5);/q;+1/p-1

InChI key

XOGTZOOQQBDUSI-UHFFFAOYSA-M

Analysis Note

Assay (iodometric): ≥ 96.0 %
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.207081.0660963705
Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200

form

powder

form

powder

form

solid

form

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

-

potency

4440 mg/kg LD50, oral (Rat)

potency

>2500 mg/kg LD50, oral (Rat)

potency

920 mg/kg LD50, oral (Rat), >10000 mg/kg LD50, skin (Rabbit)

potency

-


Still not finding the right product?

Explore all of our products under Sodium 2-mercaptoethanesulfonate


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters