Skip to Content
Merck
All Photos(1)

Key Documents

8.08514

Sigma-Aldrich

Pentanonitrile

for synthesis

Synonym(s):

Pentanonitrile, Butyl cyanide, Valeronitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€64.70

€64.70


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€64.70

About This Item

Linear Formula:
CH3(CH2)3CN
CAS Number:
Molecular Weight:
83.13
MDL number:
UNSPSC Code:
12352117
EC Index Number:
203-781-8
NACRES:
NA.22

€64.70


Estimated to ship on18 May 2025


Request a Bulk Order

vapor pressure

7.0 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

520 °C

potency

332 mg/kg LD50, oral (Rat)

bp

141 °C/1013 mbar

mp

-96 °C

transition temp

flash point 38 °C

density

0.799 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N#CCCCC

InChI

1S/C5H9N/c1-2-3-4-5-6/h2-4H2,1H3

InChI key

RFFFKMOABOFIDF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.798 - 0.800
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service