Skip to Content
Merck
All Photos(1)

Key Documents

8.08348

Sigma-Aldrich

Chlorotriphenylmethane

for synthesis

Synonym(s):

Chlorotriphenylmethane, Triphenylchloromethane, Triphenylmethyl chloride, Trityl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€90.70
1 KG
€255.00

€90.70


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€90.70
1 KG
€255.00

About This Item

Linear Formula:
ClC(C6H5)3
CAS Number:
Molecular Weight:
278.78
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-986-4
NACRES:
NA.22

€90.70


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

form

powder

potency

>5000 mg/kg LD50, oral (Rat)

bp

230-235 °C/27 hPa

mp

109-112 °C

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

ClC(c3ccccc3)(c2ccccc2)c1ccccc1

InChI

1S/C19H15Cl/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

JBWKIWSBJXDJDT-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service