Skip to Content
Merck
All Photos(1)

Key Documents

8.08272

Sigma-Aldrich

1,3-Diaminopropane

for synthesis

Synonym(s):

1,3-Diaminopropane, 1,3-Propanediamine, Trimethylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€31.70
500 ML
€50.60
7.5 KG
€562.00
45 KG
€2,670.00

€31.70


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€31.70
500 ML
€50.60
7.5 KG
€562.00
45 KG
€2,670.00

About This Item

Linear Formula:
H2N(CH2)3NH2
CAS Number:
Molecular Weight:
74.12
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-702-7
NACRES:
NA.22

€31.70


Estimated to ship on16 April 2025


Request a Bulk Order

vapor pressure

<10 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

350 °C

potency

700 mg/kg LD50, oral (Rat)
200 mg/kg LD50, skin (Rabbit)

expl. lim.

2.8-15.2 % (v/v)

pH

>12 (20 °C, 100 g/L in H2O)

bp

137-140 °C/1013 mbar

mp

-12 °C

transition temp

flash point 50 °C

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NCCCN

InChI

1S/C3H10N2/c4-2-1-3-5/h1-5H2

InChI key

XFNJVJPLKCPIBV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.886 - 0.888
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service