Skip to Content
Merck
All Photos(1)

Key Documents

8.07502

Sigma-Aldrich

3-Pentanol

for synthesis

Synonym(s):

3-Pentanol, Diethyl carbinol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€67.20
500 ML
€193.00

€67.20


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€67.20
500 ML
€193.00

About This Item

Linear Formula:
(C2H5)2CHOH
CAS Number:
Molecular Weight:
88.15
MDL number:
UNSPSC Code:
12352104
EC Index Number:
209-526-7
NACRES:
NA.22

€67.20


Estimated to ship on25 May 2025


Request a Bulk Order

vapor pressure

11.69 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

360 °C

potency

1870 mg/kg LD50, oral (Rat)
2066 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-8 % (v/v)

bp

115-116 °C/1013 hPa

mp

-8 °C

transition temp

flash point 33 °C

solubility

55 g/L

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CC)CC

InChI

1S/C5H12O/c1-3-5(6)4-2/h5-6H,3-4H2,1-2H3

InChI key

AQIXEPGDORPWBJ-UHFFFAOYSA-N

Application

3-Pentanol for synthesis. CAS No. 584-02-1, EC Number 209-526-7.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.820 - 0.821
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service