Skip to Content
Merck

Skip To

8.07477

Propylamine

for synthesis

Synonym(s):

Propylamine, 1-Aminopropane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Check Cart for Availability
€ 27,30
500 mL
Check Cart for Availability
€ 37,20
1 L
Check Cart for Availability
€ 60,30

About This Item

Empirical Formula (Hill Notation):
C3H9N
CAS Number:
Molecular Weight:
59.11
UNSPSC Code:
12352116
EC Index Number:
203-462-3
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (GC)
Form:
liquid

€ 27,30


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

330 hPa ( 20 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

325 °C

potency

370 mg/kg LD50, oral (Rat), 400 mg/kg LD50, skin (Rabbit)

expl. lim.

2.1-13.6 % (v/v)

pH

12.6 (20 °C, 100 g/L in H2O)

mp

-83 °C

transition temp

flash point <-35 °C

density

0.72 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NCCC

InChI

1S/C3H9N/c1-2-3-4/h2-4H2,1H3

InChI key

WGYKZJWCGVVSQN-UHFFFAOYSA-N

Application

  • Computational study of the unimolecular and bimolecular decomposition mechanisms of propylamine: A computational analysis revealed detailed reaction mechanisms for propylamine, focusing on dehydrogenation processes which could be crucial for developing better industrial synthesis techniques (Almatarneh et al., 2020).
  • Structural transformation and tuning behavior induced by propylamine concentration in hydrogen clathrate hydrates: This study provides fundamental understanding of guest-host distributions in clathrate hydrates, which are vital for applications in gas storage and separation technologies (Park et al., 2015).
  • Molecular association of small amphiphiles: Origin of ionic liquid properties in dibutyl phosphate/propylamine binary mixtures: The study explores the dynamic properties of propylamine mixtures, which could inform the development of new materials with ionic liquid-like characteristics for various industrial applications (Liveri et al., 2018).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.716 - 0.717
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.074768.141008.18550
assay

≥99.0% (GC)

assay

≥99.0% (GC)

assay

≥98.0% (GC)

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

density

0.72 g/cm3 at 20 °C

density

0.69 g/cm3 at 20 °C

density

0.82 g/cm3 at 20 °C

density

1.02 g/cm3 at 20 °C

mp

-83 °C

mp

-

mp

-60 °C

mp

-

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C


Still not finding the right product?

Explore all of our products under Propylamine


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

-31.0 °F

flash_point_c

< -35 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters