Skip to Content
Merck
All Photos(1)

Key Documents

8.07050

Sigma-Aldrich

2-Propyn-1-ol

for synthesis

Synonym(s):

2-Propyn-1-ol, Propargyl alcohol, Propinol, Ethynyl carbinol

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€44.60
1 L
€101.00

€44.60


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€44.60
1 L
€101.00

About This Item

CAS Number:
Molecular Weight:
56.06
MDL number:
UNSPSC Code:
12352100
EC Index Number:
203-471-2
NACRES:
NA.22

€44.60


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

10 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

365 °C

potency

56 mg/kg LD50, oral (Rat)

expl. lim.

1.9-86.2 % (v/v)

pH

3 (20 °C, 500 g/L in H2O)

bp

114-115 °C/1013 hPa

mp

-53 °C

transition temp

flash point 33 °C

solubility

1000 g/L

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCC#C

InChI

1S/C3H4O/c1-2-3-4/h1,4H,3H2

InChI key

TVDSBUOJIPERQY-UHFFFAOYSA-N

General description

for synthesis

Application

2-Propyn-1-ol for synthesis. CAS 107-19-7, pH 3 (500 g/l, H₂O, 20 °C).

Analysis Note

Assay (GC, area%)≥ 99.0 % (a/a)Density (d 20 °C/ 4 °C)0.948 - 0.950Identity (IR)passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service