Skip to Content
Merck
All Photos(1)

Key Documents

8.06753

Sigma-Aldrich

4-Pyridinecarboxylic acid hydrazide

for synthesis

Synonym(s):

4-Pyridinecarboxylic acid hydrazide, Isoniazide, 4-Pyridinecarbohydrazide, Isonicotinic acid hydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€52.80

€52.80


Estimated to ship on19 May 2025



Select a Size

Change View
100 G
€52.80

About This Item

Linear Formula:
4-(H2NNHCO)C5H4N
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352127
EC Index Number:
200-214-6
NACRES:
NA.22

€52.80


Estimated to ship on19 May 2025


Quality Level

potency

1250 mg/kg LD50, oral (Rat)

pH

6-8 (20 °C, 50 g/L in H2O)

mp

169-172 °C

transition temp

flash point >250 °C

solubility

125 g/L

bulk density

800 kg/m3

storage temp.

15-25°C

SMILES string

N(N)C(=O)c1ccncc1

InChI

1S/C6H7N3O/c7-9-6(10)5-1-3-8-4-2-5/h1-4H,7H2,(H,9,10)

InChI key

QRXWMOHMRWLFEY-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Melting range (lower value): ≥ 169 °C
Melting range (upper value): ≤ 172 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service