Skip to Content
Merck
All Photos(1)

Key Documents

8.06538

Sigma-Aldrich

Sodium methylate

for synthesis

Synonym(s):

Sodium methylate, Sodium methoxide, Sodium methanolate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€31.60
250 G
€71.40
500 G
€76.20

€31.60


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€31.60
250 G
€71.40
500 G
€76.20

About This Item

Empirical Formula (Hill Notation):
CH3NaO
CAS Number:
Molecular Weight:
54.02
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-699-5
NACRES:
NA.22

€31.60


Estimated to ship on28 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

Assay

≥97.0% (acidimetric)

form

powder

autoignition temp.

240 °C

potency

2037 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

mp

127 °C (decomposition)

density

1.3 g/cm3 at 20 °C

bulk density

500‑600 kg/m3

storage temp.

2-8°C

SMILES string

[Na+].[O-]C

InChI

1S/CH3O.Na/c1-2;/h1H3;/q-1;+1

InChI key

WQDUMFSSJAZKTM-UHFFFAOYSA-N

Application

  • Synthesis and Biological Evaluations: A study on the cyclization of berberine and magnolol with metformin in the presence of sodium methylate, exploring pharmaceutical applications. (Cao et al., 2017).
  • Synthesis of 5-Acetyl Derivatives of Alkyl 2-Furoates: This research focuses on the synthesis of acetyl derivatives using sodium methylate, demonstrating its role in organic synthesis. (Kuticheva et al., 2015).

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Assay (NaOH): ≤ 1.0 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Sol. 1 - Met. Corr. 1 - Self-heat. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service