Skip to Content
Merck
All Photos(1)

Key Documents

8.06127

Sigma-Aldrich

Morpholine

for synthesis

Synonym(s):

Morpholine, Tetrahydro-1,4-oxazine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€25.00
500 ML
€45.00
1 L
€64.40
2.5 L
€147.00
25 L
€479.00

€25.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€25.00
500 ML
€45.00
1 L
€64.40
2.5 L
€147.00
25 L
€479.00

About This Item

Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-815-1
NACRES:
NA.22

€25.00


Estimated to ship on02 June 2025


Request a Bulk Order

vapor pressure

11 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

275 °C

potency

1900 mg/kg LD50, oral (Rat)
500 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-15.2 % (v/v)

pH

11 ( in H2O, undiluted)

kinematic viscosity

2.2 cSt(20 °C)

bp

129 °C/1013 hPa

mp

-5 °C

transition temp

flash point 32 °C

density

1.00 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1CCOCC1

InChI

1S/C4H9NO/c1-3-6-4-2-5-1/h5H,1-4H2

InChI key

YNAVUWVOSKDBBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.999 - 1.001
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service