Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

Merck

Key Documents

Skip To

8.05804

4-Methyl-2-pentanol

for synthesis

Synonym(s):

4-Methyl-2-pentanol, Methyl isobutyl carbinol, Isobutyl methyl carbinol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Check Cart for Availability
€ 29,20
1 L
Check Cart for Availability
€ 68,10

About This Item

Linear Formula:
(CH3)2CHCH2CH(OH)CH3
CAS Number:
Molecular Weight:
102.17
UNSPSC Code:
12352104
EC Index Number:
203-551-7
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid

€ 29,20


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

7.06 hPa ( 25 °C)

Quality Segment

assay

≥98.0% (GC)

form

liquid

autoignition temp.

305 °C

potency

2590 mg/kg LD50, oral (Rat), 2884 mg/kg LD50, skin (Rabbit)

expl. lim.

1-5.5 % (v/v)

bp

130-133 °C/1013 hPa

mp

-60 °C

transition temp

flash point 41 °C

solubility

16.4 g/L

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CC(C)C)C

InChI

1S/C6H14O/c1-5(2)4-6(3)7/h5-7H,4H2,1-3H3

InChI key

WVYWICLMDOOCFB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.806 - 0.808
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 4-Methyl-2-pentanol


pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

105.8 °F - closed cup

flash_point_c

41 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

108-11-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters