Skip to Content
Merck
All Photos(1)

Key Documents

8.05791

Sigma-Aldrich

Mesityl oxide

for synthesis

Synonym(s):

Mesityl oxide, Isopropylideneacetone, 4-Methyl-3-penten-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€25.70
1 L
€144.00

€25.70


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€25.70
1 L
€144.00

About This Item

Linear Formula:
(CH3)2CCHCOCH3
CAS Number:
Molecular Weight:
98.14
MDL number:
UNSPSC Code:
12352115
EC Index Number:
205-502-5
NACRES:
NA.22

€25.70


Estimated to ship on02 June 2025


Request a Bulk Order

vapor pressure

10.5 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

344 °C

potency

1120 mg/kg LD50, oral (Rat)
5150 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-10.1 % (v/v)

bp

130 °C/1013 hPa

mp

-59 °C

transition temp

flash point 28 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(C)C=C(C)C

InChI

1S/C6H10O/c1-5(2)4-6(3)7/h4H,1-3H3

InChI key

SHOJXDKTYKFBRD-UHFFFAOYSA-N

Application

  • Nanomaterial synthesis: Mesityl oxide is employed in the chemical reactivity studies of the tryptophan/acetone/DMSO triad system, highlighting its potential applications in the innovative synthesis of nanomaterials, advancing green chemistry initiatives (Huang et al., 2023).
  • Electrode modification for residue detection: Utilized in the detection of residual 2-Phenylphenol on lemon rind, mesityl oxide served as a solvent in the electrochemical deposition of polymeric stackings, illustrating its utility in enhancing analytical methods in food safety (Kiss et al., 2022).
  • Organic sensing media: Mesityl oxide was a key solvent in the anodic polymerization process for sensing phenols, indicating its effective use in the development of sensing layers for environmental monitoring (Kiss et al., 2022).
  • Pharmaceutical product quality control: Mesityl oxide is instrumental in the determination of acetone and related impurities in drug product intermediates, showing its critical role in ensuring pharmaceutical quality and compliance (Quirk et al., 2014).

Analysis Note

Assay isomers (GC, area%): ≥ 97.0 % (a/a)
a-Mesityl oxide (GC, area%): ≥ 85.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.853 - 0.856
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service