Skip to Content
Merck
All Photos(1)

Key Documents

8.04778

Sigma-Aldrich

Ionone

for synthesis

Synonym(s):

Ionone

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€44.60

€44.60


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€44.60

About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
MDL number:
UNSPSC Code:
12352115
EC Index Number:
232-396-8
NACRES:
NA.22

€44.60


Estimated to ship on21 May 2025


Request a Bulk Order

vapor pressure

0.0013 hPa ( 20 °C)

Quality Level

form

liquid

potency

4590 mg/kg LD50, oral (Rat)

bp

237 °C/1013 hPa

transition temp

flash point 117 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(C)\C=C\C1C(CCC=C1C)(C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h6-8,12H,5,9H2,1-4H3/b8-7+

InChI key

UZFLPKAIBPNNCA-BQYQJAHWSA-N

Application

Ionone for synthesis. CAS 8013-90-9, molar mass 192.3 g/mol.

Analysis Note

Sum α- und β – Isomers (GC): ≥ 90.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.929 - 0.935
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

242.6 °F

Flash Point(C)

117 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service