Skip to Content
Merck
All Photos(1)

Key Documents

8.04536

Sigma-Aldrich

4-Hydroxybenzaldehyde

for synthesis

Synonym(s):

4-Hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€53.10
250 G
€102.00
5 KG
€685.00

€53.10


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€53.10
250 G
€102.00
5 KG
€685.00

About This Item

Linear Formula:
4-(OH)C6H4CHO
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352114
EC Index Number:
204-599-1
NACRES:
NA.22

€53.10


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

autoignition temp.

450 °C

potency

2250 mg/kg LD50, oral (Rat)

mp

116 °C

transition temp

flash point 174 °C

solubility

13.8 g/L

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Oc1ccc(cc1)C=O

InChI

1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H

InChI key

RGHHSNMVTDWUBI-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98 %
Melting range
- lower value:
≥ 114 °C
- upper value: ≤ 118 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

345.2 °F

Flash Point(C)

174 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service