Skip to Content
Merck
All Photos(1)

Key Documents

8.04079

Sigma-Aldrich

Germanium(IV) chloride

for synthesis

Synonym(s):

Germanium(IV) chloride, Germanium tetrachloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€488.00

€488.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€488.00

About This Item

Empirical Formula (Hill Notation):
Cl4Ge
CAS Number:
Molecular Weight:
214.45
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-116-7
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
liquid

€488.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

101 hPa

form

liquid

pH

<1 (20 °C in H2O)

bp

83.1 °C

mp

-49.5 °C

density

1.88 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/4Cl.Ge

InChI key

LBXSGGYKZPEYRS-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (argentometric): ≥ 99.0 %
Density (d 20 °C/ 4 °C): 1.875 - 1.877
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Does not sustain combustion.

Flash Point(C)

Does not sustain combustion.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service