Skip to Content
Merck
All Photos(1)

Documents

8.03978

Sigma-Aldrich

Ferrocene

for synthesis

Synonym(s):

Ferrocene, Dicyclopentadienyliron, Iron dicyclopentadienyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C5H5)2Fe
CAS Number:
Molecular Weight:
186.03
MDL number:
UNSPSC Code:
12352103
EC Index Number:
203-039-3
NACRES:
NA.22

vapor pressure

3.4 hPa ( 100 °C)

Quality Level

Assay

≥98.0% (GC)

form

crystals

autoignition temp.

>150 °C

potency

1320 mg/kg LD50, oral (Rat)
>3000 mg/kg LD50, skin (Rat)

density

1.49 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

2-30°C

InChI

1S/2C5H5.Fe/c2*1-2-4-5-3-1;/h2*1-5H;/q2*-1;+2

InChI key

KTWOOEGAPBSYNW-UHFFFAOYSA-N

Application

  • Spin and Charge Interactions between Nanographene Host and Ferrocene: This study investigates the charge transfer mechanisms between ferrocene molecules and carbon nanotubes (CNTs), exploring magnetic interactions and potential applications in electronic devices (A Suzuki et al., 2024).
  • Development of Novel Immobilized Copper–Ligand Complex for Click Chemistry of Biomolecules: Research on coumarin-ferrocene triazoles for potential use in biochemical applications, highlighting small molecule interactions and novel click chemistry approaches (R Kandler et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 1 - Flam. Sol. 1 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Liver

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service