Skip to Content
Merck
All Photos(1)

Key Documents

8.03771

Sigma-Aldrich

2,5-Dimethylphenol

for synthesis

Synonym(s):

2,5-Dimethylphenol, 2,5-DMP, p-Xylenol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€25.00
250 G
€90.10

€25.00


Estimated to ship on25 May 2025



Select a Size

Change View
5 G
€25.00
250 G
€90.10

About This Item

Linear Formula:
2,5-(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-461-5
NACRES:
NA.22

€25.00


Estimated to ship on25 May 2025


vapor pressure

0.208 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

expl. lim.

1.4 % (v/v)

mp

75 °C

transition temp

flash point 80-95 °C

solubility

slightly soluble 3.54 g/L

density

0.9 g/cm3 at 75 °C

storage temp.

2-30°C

SMILES string

Oc1c(ccc(c1)C)C

InChI

1S/C8H10O/c1-6-3-4-7(2)8(9)5-6/h3-5,9H,1-2H3

InChI key

NKTOLZVEWDHZMU-UHFFFAOYSA-N

Application

2,5-Dimethylphenol for synthesis. CAS 95-87-4, chemical formula 2,5-(CH₃)₂C₆H₃OH.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 73 °C
Melting range (upper value): ≤ 76 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service