Skip to Content
Merck
All Photos(2)

Key Documents

8.03603

Sigma-Aldrich

1,5-Diazabicyclo[4.3.0]non-5-ene

for synthesis

Synonym(s):

1,5-Diazabicyclo[4.3.0]non-5-ene, Pyrrolidino[1,2:a]1,4,5,6-tetrahydropyrimidine, DBN

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€93.20
100 ML
€241.00

€93.20


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€93.20
100 ML
€241.00

About This Item

Empirical Formula (Hill Notation):
C7H12N2
CAS Number:
Molecular Weight:
124.18
MDL number:
UNSPSC Code:
12352005
EC Index Number:
221-087-3
NACRES:
NA.22

€93.20


Estimated to ship on26 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

Assay

≥97.0% (GC)

form

liquid

bp

110 °C/20 hPa

transition temp

flash point 94 °C

density

1.04 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H12N2/c1-3-7-8-4-2-6-9(7)5-1/h1-6H2

InChI key

SGUVLZREKBPKCE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.038 - 1.041
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service