Skip to Content
Merck
All Photos(1)

Key Documents

8.03513

Sigma-Aldrich

Diphenylmethane

for synthesis

Synonym(s):

Diphenylmethane, Benzylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CH2
CAS Number:
Molecular Weight:
168.23
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-978-6
form:
crystals
Assay:
≥98.5% (GC)

vapor pressure

1 hPa ( 77 °C)

Assay

≥98.5% (GC)

form

crystals

autoignition temp.

485 °C

potency

2250 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.69-8.66 % (v/v)

bp

264-266 °C/1013 hPa

mp

22-25 °C

transition temp

flash point 130 °C

solubility

0.014 g/L

bulk density

1006 kg/m3

storage temp.

2-30°C

InChI

1S/C13H12/c1-3-7-12(8-4-1)11-13-9-5-2-6-10-13/h1-10H,11H2

InChI key

CZZYITDELCSZES-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.5 % (a/a)
Melting range (lower value): ≥ 22 °C
Melting range (upper value): ≤ 25 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(C)

130 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service