Skip to Content
Merck
All Photos(2)

Key Documents

8.03273

Sigma-Aldrich

2-(Diethylamino)-ethanol

for synthesis

Synonym(s):

2-(Diethylamino)-ethanol, N,N,-Diethylethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€34.10
1 L
€47.10
25 L
€445.00

€34.10


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
100 ML
€34.10
1 L
€47.10
25 L
€445.00

About This Item

Linear Formula:
(C2H5)2NCH2CH2OH
CAS Number:
Molecular Weight:
117.19
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-845-2
NACRES:
NA.22

€34.10


Only 0 left in stock

Request a Bulk Order

vapor pressure

1.9 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

270 °C

potency

1300 mg/kg LD50, oral (Rat)
1109 mg/kg LD50, skin (Rabbit)

expl. lim.

0.7 % (v/v)

pH

11.5 (20 °C, 100 g/L in H2O)

bp

163 °C/1013 hPa

mp

-68 °C

transition temp

flash point 51 °C

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCO)(CC)CC

InChI

1S/C6H15NO/c1-3-7(4-2)5-6-8/h8H,3-6H2,1-2H3

InChI key

BFSVOASYOCHEOV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.883 - 0.885
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service