Skip to Content
Merck
All Photos(4)

Key Documents

8.02898

Sigma-Aldrich

Diethyl carbonate

greener alternative

for synthesis

Synonym(s):

Diethyl carbonate, Carbonic acid diethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€26.60
500 ML
€38.50
1 L
€49.10

€26.60


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€26.60
500 ML
€38.50
1 L
€49.10

About This Item

Linear Formula:
CO(OC2H5)2
CAS Number:
Molecular Weight:
118.13
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-311-1
NACRES:
NA.22

€26.60


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

11 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

445 °C

potency

>4900 mg/kg LD50, oral (Rat)

expl. lim.

1.4-11.0 % (v/v)

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

6.3 (1 g/L in H2O)

bp

126 °C/1013 hPa

mp

-43 °C

transition temp

flash point 33 °C

solubility

insoluble 18.8 mg/L

density

0.97 g/cm3 at 20 °C

greener alternative category

storage temp.

no temp limit

SMILES string

O(CC)C(=O)OCC

InChI

1S/C5H10O3/c1-3-7-5(6)8-4-2/h3-4H2,1-2H3

InChI key

OIFBSDVPJOWBCH-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is used as a greener reagent. Click here for more information.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.974 - 0.975
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Green chemicals evaluation aligns with the 12 Principles of Green Chemistry, emphasizing resource efficiency and hazard reduction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service