Skip to Content
Merck
All Photos(1)

Key Documents

8.02885

Sigma-Aldrich

Cyclohexylamine

for synthesis

Synonym(s):

Cyclohexylamine, Aminocyclohexane, Cyclohexanamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€23.40
1 L
€37.70
2.5 L
€76.20

€23.40


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€23.40
1 L
€37.70
2.5 L
€76.20

About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-629-0
NACRES:
NA.22

€23.40


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

14 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

265 °C

potency

300 mg/kg LD50, oral (Rat)

expl. lim.

1.6-9.4 % (v/v)

pH

11.5 (20 °C, 100 g/L in H2O)

bp

133-134 °C/1013 hPa

mp

-18 °C

transition temp

flash point 27 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NC1CCCCC1

InChI

1S/C6H13N/c7-6-4-2-1-3-5-6/h6H,1-5,7H2

InChI key

PAFZNILMFXTMIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Fluorescent sensor for aliphatic amines: Research on a mixed-linkage donor-acceptor covalent organic framework reveals its potential as a turn-on fluorescent sensor for aliphatic amines, including cyclohexylamine, providing insights into its applications in chemical sensing technologies (Yue et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.866 - 0.868
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service