Skip to Content
Merck
All Photos(1)

Key Documents

8.02835

Sigma-Aldrich

Cysteaminium chloride

for synthesis

Synonym(s):

Cysteaminium chloride, 2-Aminoethanethiolhydrochloride, Cysteamine hydrochloride, 2-Mercaptoethylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€36.00
25 G
€43.00
100 G
€125.00

€36.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
€36.00
25 G
€43.00
100 G
€125.00

About This Item

Empirical Formula (Hill Notation):
C2H7NS · xHCl
CAS Number:
Molecular Weight:
77.15 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-858-1
NACRES:
NA.22

€36.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

form

crystals

potency

642 mg/kg LD50, oral (Rat)

pH

3.3-5.0 (20 °C, 400 g/L in H2O)

mp

66-69 °C

solubility

400 g/L

storage temp.

2-8°C

SMILES string

SCC[N+H3].[Cl-]

InChI

1S/C2H7NS.ClH/c3-1-2-4;/h4H,1-3H2;1H

InChI key

OGMADIBCHLQMIP-UHFFFAOYSA-N

Application

  • Synthesis of nanoparticles: A study on the functionalization of multiwalled carbon nanotubes using Cysteaminium chloride was conducted, resulting in the synthesis of gold nanoparticles decorated carbon nanotubes. This highlights its application in nanomaterials fabrication and potential uses in various technological fields (Duc Chinh et al., 2019).
  • Pharmaceutical applications: Cysteaminium chloride′s chemical properties are utilized in the synthesis of thiol-functionalized chitosan nanoparticles. These nanoparticles are designed to adsorb heavy metals from aqueous solutions, demonstrating a vital application in environmental management and potential for pharmaceutical waste treatment (Nemati et al., 2019).
  • Crystallography: Research involving Cysteaminium chloride has contributed to the discovery of a monoclinic polymorph of cysteamine hydrochloride, which is significant for its implications in crystal engineering and materials science, providing insights into molecular and crystal structure interactions (Ahmad et al., 2009).

Analysis Note

Assay (iodometric): ≥ 97.0 %
Melting range (lower value): ≥ 61 °C
Melting range (upper value): ≤ 67 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service