Skip to Content
Merck

Skip To

8.01966

2-Bromo-2-methylpropane

(stabilised with potassium carbonate) for synthesis

Synonym(s):

2-Bromo-2-methylpropane, tert-Butyl bromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mL
Check Cart for Availability
€ 71,30
1 L
Check Cart for Availability
€ 260,00

About This Item

Linear Formula:
(CH3)3CBr
CAS Number:
Molecular Weight:
137.02
UNSPSC Code:
12352101
EC Index Number:
208-065-9
NACRES:
NA.22
MDL number:
Assay:
≥97.0% (GC)
Form:
liquid

€ 71,30


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

179.9 hPa ( 25 °C)

Quality Segment

assay

≥97.0% (GC)

form

liquid

mp

-20 °C

transition temp

flash point 16 °C

density

1.22 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrC(C)(C)C

InChI

1S/C4H9Br/c1-4(2,3)5/h1-3H3

InChI key

RKSOPLXZQNSWAS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.215 - 1.225
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.01550325066135615
assay

≥97.0% (GC)

assay

≥99% (GC)

assay

97%

assay

98%

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

mp

-20 °C

mp

-28 °C

mp

-

mp

−20 °C (lit.)

storage temp.

2-30°C

storage temp.

-

storage temp.

-

storage temp.

-

density

1.22 g/cm3 at 20 °C

density

0.84 g/cm3 at 20 °C

density

1.461 g/mL at 20 °C (lit.)

density

1.22 g/mL at 20 °C (lit.)

form

liquid

form

liquid

form

liquid

form

liquid


Still not finding the right product?

Explore all of our products under 2-Bromo-2-methylpropane


pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

60.8 °F - closed cup

flash_point_c

16 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters