Skip to Content
Merck
All Photos(1)

Documents

8.01636

Sigma-Aldrich

Ethyl bromoacetate

for synthesis

Synonym(s):

Ethyl bromoacetate, Bromoacetic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7BrO2
CAS Number:
Molecular Weight:
167.00
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-290-9
NACRES:
NA.22

vapor pressure

4.5 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

158-161 °C/1013 hPa

mp

-38 °C

transition temp

flash point 50 °C

density

1.50 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H7BrO2/c1-2-7-4(6)3-5/h2-3H2,1H3

InChI key

PQJJJMRNHATNKG-UHFFFAOYSA-N

Application

  • Synthesis and anticancer potential of thiazolidinone and thiadiazole derivatives: Ethyl bromoacetate was utilized in the synthesis of new thiazolidinone and thiadiazole derivatives, which were subsequently evaluated for their anticancer properties through molecular docking studies. This application highlights its role in the development of potential anticancer agents (Saeed et al., 2024).
  • Development of pyrazolopyranopyrimidine derivatives: Research on the synthesis of novel pyrazolopyranopyrimidine derivatives involved ethyl bromoacetate, focusing on their structural and dynamic properties using crystallography, DFT calculations, and molecular dynamics simulations, contributing to the field of medicinal chemistry (Ait Elmachkouri et al., 2023).
  • Eco-friendly insecticidal derivatives for olive pest control: Ethyl bromoacetate was employed in the design and synthesis of pyridine and thiazole derivatives aimed at providing eco-friendly insecticidal solutions for controlling olive pests, demonstrating its utility in agricultural chemistry (Elkanzi et al., 2023).
  • DNA-compatible synthesis of thiazolidione derivatives: A study used ethyl bromoacetate in a DNA-compatible three-component annulation and Knoevenagel condensation process to synthesize thiazolidione derivatives, showcasing its versatility in complex organic syntheses (Wang et al., 2023).
  • Accidental discovery of macrocycles: Ethyl bromoacetate played a role in the serendipitous discovery of new macrocycles containing amide, amino, and carbamate groups during a residue separation process, illustrating its potential in unexpected synthetic outcomes (Horita et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.500 - 1.510
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service