Skip to Content
Merck

Skip To

8.01464

2-Amino-2-methyl-1,3-propanediol

for synthesis

Synonym(s):

2-Amino-2-methyl-1,3-propanediol, Ammediol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H2N(CH3)C(CH2OH)2
CAS Number:
Molecular Weight:
105.14
UNSPSC Code:
12352104
EC Index Number:
204-100-7
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

13.3 hPa ( 152 °C)

Quality Segment

form

solid

potency

17000 mg/kg LD50, oral (Rat)

pH

10-11 (20 °C, 10 g/L in H2O)

bp

150-152 °C/13 hPa

mp

102-105 °C

transition temp

flash point >100 °C

storage temp.

2-30°C

SMILES string

NC(CO)(CO)C

InChI

1S/C4H11NO2/c1-4(5,2-6)3-7/h6-7H,2-3,5H2,1H3

InChI key

UXFQFBNBSPQBJW-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
085698.18522A9754
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

form

solid

form

crystalline

form

liquid

form

crystalline

pH

10-11 (20 °C, 10 g/L in H2O)

pH

10.5-11.5 (25 °C, 1 M in H2O)

pH

11 (20 °C, 100 g/L in H2O)

pH

10.5-11.5 (25 °C, 105.1 g/L)

storage temp.

2-30°C

storage temp.

-

storage temp.

2-30°C

storage temp.

-

mp

102-105 °C

mp

100-110 °C (lit.)

mp

-

mp

100-110 °C (lit.)

bp

150-152 °C/13 hPa

bp

151 °C/10 mmHg (lit.)

bp

-

bp

151 °C/10 mmHg (lit.)


Still not finding the right product?

Explore all of our products under 2-Amino-2-methyl-1,3-propanediol


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

>212.0 °F

flash_point_c

> 100 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters