Skip to Content
Merck
All Photos(1)

Key Documents

8.01417

Sigma-Aldrich

1-Aminoguanidinium hydrogen carbonate

for synthesis

Synonym(s):

1-Aminoguanidinium hydrogen carbonate, Aminoguanidine bicarbonate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€39.40
25 KG
€702.00

€39.40


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€39.40
25 KG
€702.00

About This Item

Empirical Formula (Hill Notation):
C2H8N4O3
CAS Number:
Molecular Weight:
136.11
MDL number:
UNSPSC Code:
12352107
EC Index Number:
219-956-7
NACRES:
NA.22

€39.40


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

form

powder

autoignition temp.

245 °C

potency

>5000 mg/kg LD50, oral (Rat)

pH

8.9 (20 °C, 5 g/L in H2O)

mp

125 °C (decomposes)

solubility

3.3 g/L

density

1.56 g/cm3 at 20 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

[N+H2](N)\C(=N\[H])\N.[O-]C(=O)O

InChI

1S/CH6N4.CH2O3/c2-1(3)5-4;2-1(3)4/h4H2,(H4,2,3,5);(H2,2,3,4)

InChI key

OTXHZHQQWQTQMW-UHFFFAOYSA-N

Application

  • The chemistry of aminoguanidine derivatives – preparation, crystal structure, thermal properties, and molecular docking studies of aminoguanidinium salts: Explores the preparation and properties of aminoguanidine derivatives, highlighting the versatility of aminoguanidinium salts in various chemical contexts (R Selvakumar et al., 2015).

Analysis Note

Color (visual): white to off white
Assay (HClO₄): ≥ 97.0 %
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service