Skip to Content
Merck
All Photos(1)

Key Documents

8.00885

Sigma-Aldrich

Trimethyl orthoformate

for synthesis

Synonym(s):

Trimethyl orthoformate, Methyl orthoformate, Orthoformic acid trimethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€31.30
1 L
€72.50

€31.30


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€31.30
1 L
€72.50

About This Item

Linear Formula:
CH(OCH3)3
CAS Number:
Molecular Weight:
106.12
MDL number:
UNSPSC Code:
12352112
EC Index Number:
205-745-7
NACRES:
NA.22

€31.30


Estimated to ship on01 June 2025


Request a Bulk Order

vapor pressure

10 hPa ( 7 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

255 °C

potency

3130 mg/kg LD50, oral (Rat)

expl. lim.

1.4-44.6 % (v/v)

bp

102 °C/1013 hPa

mp

-53 °C

transition temp

flash point 13 °C

solubility

10 g/L (Hydrolysis)

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(OC)OC)C

InChI

1S/C4H10O3/c1-5-4(6-2)7-3/h4H,1-3H3

InChI key

PYOKUURKVVELLB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.968 - 0.970
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service