Skip to Content
Merck
All Photos(3)

Key Documents

8.00857

Sigma-Aldrich

2-Ethoxyethanol (stabilised)

for synthesis

Synonym(s):

2-Ethoxyethanol, Ethyl glycol, Ethylene glycol monoethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1098271
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-804-1
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
135 °C (lit.)
vapor pressure:
3.8 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

vapor density

3.1 (vs air)

vapor pressure

3.8 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

460 °F

potency

3300 mg/kg LD50, skin (Rabbit)

expl. lim.

14 %

dilution

(for synthesis)

refractive index

n20/D 1.407 (lit.)

pH

7 (20 °C)

bp

135 °C (lit.)

mp

−90 °C (lit.)

transition temp

flash point 40 °C

density

0.93 g/mL at 25 °C (lit.)

storage temp.

no temp limit

SMILES string

CCOCCO

InChI

1S/C4H10O2/c1-2-6-4-3-5/h5H,2-4H2,1H3

InChI key

ZNQVEEAIQZEUHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemical synthesis applications

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.929 - 0.930
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test
The material is stabilised with 2,6-Di-tert-butyl-4-methylphenol. At time of analytical testing a content of >10 ppm stabiliser was determined.
The material is stabilised with 2,6-Di-tert-butyl-4-methylphenol. At time of analytical testing a content of >10 ppm stabiliser was determined.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service