Skip to Content
Merck
All Photos(1)

Key Documents

8.00813

Sigma-Aldrich

Undecylenic-10 acid

for synthesis

Synonym(s):

Undecylenic-10 acid, 10-Undecenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€68.00
5 L
€520.00

€68.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€68.00
5 L
€520.00

About This Item

Linear Formula:
CH2=CH(CH2)8COOH
CAS Number:
Molecular Weight:
184.28
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-965-8
NACRES:
NA.22

€68.00


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

solid

autoignition temp.

275 °C

potency

2500 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

275 °C/1013 mbar

mp

24 °C

transition temp

flash point 149 °C

solubility

73.7 mg/L

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)CCCCCCCCC=C

InChI

1S/C11H20O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2H,1,3-10H2,(H,12,13)

InChI key

FRPZMMHWLSIFAZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 21 °C
Melting range (upper value): ≤ 25 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

300.2 °F

Flash Point(C)

149 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service