Skip to Content
Merck
All Photos(1)

Key Documents

8.00645

Sigma-Aldrich

3-Nitrobenzoic acid

for synthesis

Synonym(s):

3-Nitrobenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€31.50
25 KG
€615.00

€31.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€31.50
25 KG
€615.00

About This Item

Linear Formula:
3-(NO2)C6H4COOH
CAS Number:
Molecular Weight:
167.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-508-5
NACRES:
NA.22

€31.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99% (HPLC)

form

crystals

autoignition temp.

490 °C

pH

3 (20 °C, 5 g/L in H2O, aqueous suspension)

mp

139-142 °C

transition temp

flash point 190 °C

solubility

3 g/L

density

1.49 g/cm3 at 20 °C

bulk density

700 kg/m3

storage temp.

no temp limit

SMILES string

[N+](=O)([O-])c1cc(ccc1)C(=O)O

InChI

1S/C7H5NO4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H,(H,9,10)

InChI key

AFPHTEQTJZKQAQ-UHFFFAOYSA-N

Related Categories

Application

  • CO–CRYSTAL SYNTHESIS AND STRUCTURAL ANALYSIS OF PARACETAMOL AND 3–NITROBENZOIC ACID: This study focuses on the co-crystallization of paracetamol with 3-nitrobenzoic acid, providing insights into the structural dynamics of the resulting compound (PP Chahande, 2023).
  • Photocatalytic degradation of methylene blue dye and electrocatalytic water oxidation over copper (II) complex with mixed ligands: This study explores the use of a copper (II) complex with 3-nitrobenzoic acid in photocatalytic degradation and electrocatalytic water oxidation (M Jamil et al., 2024).

Analysis Note

Assay (HPLC, area%): ≥ 99 %
Melting range
- lower value:
≥ 139 °C
- upper value: ≤ 143 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

373.3 °F - Pensky-Martens closed cup

Flash Point(C)

189.6 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service