Skip to Content
Merck
All Photos(1)

Key Documents

8.00420

Sigma-Aldrich

3-Aminophenol

for synthesis

Synonym(s):

3-Aminophenol, 3-Hydroxyaniline

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€30.40
250 G
€57.00
1 KG
€153.00
50 KG
€4,050.00

€30.40


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€30.40
250 G
€57.00
1 KG
€153.00
50 KG
€4,050.00

About This Item

Linear Formula:
3-(NH2)C6H4OH
CAS Number:
Molecular Weight:
109.13
MDL number:
UNSPSC Code:
12352104
EC Index Number:
209-711-2
NACRES:
NA.22

€30.40


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

>600 °C

potency

924 mg/kg LD50, oral (Rat)

pH

6.8 (20 °C, 10 g/L in H2O)

bp

164 °C/15 hPa

mp

122 °C

transition temp

flash point 178 °C

solubility

26 g/L

density

0.99 g/cm3 at 25 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

Nc1cc(ccc1)O

InChI

1S/C6H7NO/c7-5-2-1-3-6(8)4-5/h1-4,8H,7H2

InChI key

CWLKGDAVCFYWJK-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Melting range (lower value): ≥ 120 °C
Melting range (upper value): ≤ 123 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service