Skip to Content
Merck
All Photos(1)

Key Documents

8003SCPKIT

Millipore

Large Replacement Parts for 8003 Series Stirred Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
eCl@ss:
32019190
NACRES:
NB.24

shipped in

ambient

Components

Cap assembly - Part no. 1524901 - Qty/Kit- 1
Stirrer assembly - Part no. 1570801 - Qty/Kit- 1
base - Part no. 21000 - Qty/Kit- 1

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Wang et al.
Methods in molecular biology (Clifton, N.J.), 2664, 41-53 (2023-07-10)
The glomerular basement membrane (GBM) is an important component of the kidney filtration barrier. The ability to evaluate the molecular transport properties of the GBM and determining how changes in the structure, composition, and mechanical properties of the GBM regulate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service