Skip to Content
Merck
All Photos(2)

Key Documents

8.00361

Sigma-Aldrich

2-(1-Piperazinyl)ethylamine

for synthesis

Synonym(s):

2-(1-Piperazinyl)ethylamine, N-(2-Aminoethyl)piperazine, AEP, 2-Piperazinoethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€34.30

€34.30


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€34.30

About This Item

Empirical Formula (Hill Notation):
C6H15N3
CAS Number:
Molecular Weight:
129.20
MDL number:
UNSPSC Code:
12352005
EC Index Number:
205-411-0
NACRES:
NA.22
form:
liquid
Assay:
≥98.0% (GC)

€34.30


Estimated to ship on30 March 2025


Request a Bulk Order

vapor pressure

0.07 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

315 °C

potency

1470 mg/kg LD50, oral (Rat)

expl. lim.

2.1-10.5 % (v/v)

pH

12 (20 °C, 100 g/L in H2O)

bp

222 °C/1013 hPa

mp

-19 °C

transition temp

flash point 88 °C

solubility

>1000 g/L

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1(CCNCC1)CCN

InChI

1S/C6H15N3/c7-1-4-9-5-2-8-3-6-9/h8H,1-7H2

InChI key

IMUDHTPIFIBORV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.983 - 0.987
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service