Skip to Content
Merck
All Photos(2)

Key Documents

8.00273

Sigma-Aldrich

4′-Aminoacetophenone

for synthesis

Synonym(s):

4′-Aminoacetophenone, 1-(4-Aminophenyl)-ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(NH2)C6H4COCH3
CAS Number:
Molecular Weight:
135.16
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-801-2

Assay

≥99.0% (GC)

form

powder

potency

381 mg/kg LD50, oral (Rat)

bp

292-294 °C/1013 hPa

mp

103-106 °C

solubility

6.5 g/L

bulk density

580 kg/m3

storage temp.

2-30°C

InChI

1S/C8H9NO/c1-6(10)7-2-4-8(9)5-3-7/h2-5H,9H2,1H3

InChI key

GPRYKVSEZCQIHD-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 103 °C
Melting range (upper value): ≤ 106 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service