Skip to Content
Merck
All Photos(1)

Key Documents

8.00145

Sigma-Aldrich

6-Aminohexanoic acid

for synthesis

Synonym(s):

6-Aminohexanoic acid, 6-Aminocaproic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€29.90
250 G
€51.70
1 KG
€177.00
5 KG
€856.00
10 KG
€1,390.00

€29.90


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€29.90
250 G
€51.70
1 KG
€177.00
5 KG
€856.00
10 KG
€1,390.00

About This Item

Linear Formula:
H2N(CH2)5COOH
CAS Number:
Molecular Weight:
131.17
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-469-3
NACRES:
NA.22

€29.90


Estimated to ship on28 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

<0.1 hPa ( 20 °C)

Assay

≥99% (HClO4)

form

crystals

potency

3400 mg/kg LD50, oral (Rat)

pH

6.27-6.3 (20 °C, 587.55 g/L in H2O)

mp

205 °C

transition temp

flash point 207-209 °C

solubility

587.55 g/L

density

1.1313 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[N+H3]CCCCCC(=O)[O-]

InChI

1S/C6H13NO2/c7-5-3-1-2-4-6(8)9/h1-5,7H2,(H,8,9)

InChI key

SLXKOJJOQWFEFD-UHFFFAOYSA-N

Application

  • Computational Insights for Wastewater Treatment: A study provided computational insights into the reactivity of crosslinked polyketones with 6-Aminohexanoic acid modifications, demonstrating potential for enhanced copper ion removal from wastewater, suggesting a valuable application in environmental remediation processes (Ortega et al., 2023).

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

404.6 - 408.2 °F

Flash Point(C)

207 - 209 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service