Skip to Content
Merck
All Photos(1)

Key Documents

8.00133

Sigma-Aldrich

Barbituric acid

for synthesis

Synonym(s):

Barbituric acid, 2,4,6-Trihydroxypyrimidine, N,N′-Malonylurea

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€42.90
250 G
€89.70

€42.90


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€42.90
250 G
€89.70

About This Item

Empirical Formula (Hill Notation):
C4H4N2O3
CAS Number:
Molecular Weight:
128.09
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-658-0
NACRES:
NA.22

€42.90


Estimated to ship on24 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

Assay

≥99% (acidimetric)

form

solid

potency

>5000 mg/kg LD50, oral (Rat)

pH

2-3 (60 °C, 50 g/L in H2O)

mp

250-252 °C (decomposition)

solubility

11.45 g/L

bulk density

720 kg/m3

storage temp.

2-30°C

InChI

1S/C4H4N2O3/c7-2-1-3(8)6-4(9)5-2/h1H2,(H2,5,6,7,8,9)

InChI key

HNYOPLTXPVRDBG-UHFFFAOYSA-N

Application

  • Synthesis of pyrazolopyranopyrimidines: A study employed a Co-MOF@Ag(2)O nanocomposite as a nano-organic catalyst for the efficient one-pot synthesis of pyrazolopyranopyrimidines, highlighting its potential application in organic synthesis involving barbituric acid derivatives (Hootifard et al., 2023).
  • Optical and electrochemical characterization of organic dyes: Research on the optical, electrochemical, thermal, and theoretical properties of carbazole-derived organic dyes noted implications for the synthesis methods and applications of barbituric acid in dye chemistry, indicating advancements in material science (Naik et al., 2024).

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service