Skip to Content
Merck
All Photos(2)

Key Documents

8.00023

Sigma-Aldrich

Acetylacetone

for synthesis

Synonym(s):

ACAC, 2,4-Pentanedione

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€27.20
250 ML
€34.10
1 L
€85.00
10 L
€472.00

€27.20


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€27.20
250 ML
€34.10
1 L
€85.00
10 L
€472.00

About This Item

Linear Formula:
CH3COCH2COCH3
CAS Number:
Molecular Weight:
100.12
Beilstein:
741937
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-634-0
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
140.4 °C (lit.)
vapor pressure:
6 mmHg ( 20 °C)

€27.20


Estimated to ship on01 June 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor density

3.5 (vs air)

vapor pressure

6 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

662 °F

potency

575 mg/kg LD50, oral (Rat)

expl. lim.

11.4 %

dilution

(for synthesis)

refractive index

n20/D 1.452 (lit.)

pH

6 (20 °C, 200 g/L in H2O)

bp

140.4 °C (lit.)

mp

−23 °C (lit.)

transition temp

flash point 35 °C

solubility

153.8-155.2 g/L

density

0.975 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(=O)CC(C)=O

InChI

1S/C5H8O2/c1-4(6)3-5(2)7/h3H2,1-2H3

InChI key

YRKCREAYFQTBPV-UHFFFAOYSA-N

Application

  • Viscoelasticity in Pressure-Sensitive Adhesives: This study examines the influence of crosslink density and linear viscoelasticity on the shear failure of adhesives, showcasing Aluminium acetylacetonate′s role in enhancing the performance and durability of industrial adhesives (Arrowood et al., 2023).
  • Heat Stabilizer in PVC: Discusses the preparation of magnesium-aluminum hydrotalcite using a mechanochemical method facilitated by Aluminium acetylacetonate, and its application as a heat stabilizer in poly(vinyl chloride), highlighting its importance in improving the thermal stability of polymers (Jiang et al., 2020).

Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 0.972 - 0.974
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service