Skip to Content
Merck

Skip To

79146BC

GDS PickPen® PIPETMAX® Surface Keyboard Cover

BioControl Systems, for use with GDS PPMX Microsoft Surface Laptop

Synonym(s):

GDS PPMX Surface Keyboard Cvr, Surface Keyboard Case

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 257,00

About This Item

UNSPSC Code:
41121521
NACRES:
NC.81

€ 257,00


Check Cart for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages

compatibility

for use with GDS PPMX Microsoft Surface Laptop

storage temp.

room temp

General description

GDS PickPen® PIPETMAX® (PPMX) Surface Keyboard Cover is a detachable keyboard accessory designed for Microsoft Surface devices. It serves as a protective cover when closed and can be flipped open to provide a full-size keyboard for typing. The Surface Keyboard Cover is wireless and connects to the Surface device magnetically, allowing easy pairing and a seamless typing experience. It is a popular accessory for Surface tablets and laptops, providing a convenient way to enhance productivity while on the go.

Application

GDS PPMX Surface Keyboard Cover is used with GDS PickPen® PIPETMAX® Surface laptop to complete sample preparation for the detection of pathogenic organisms in foods, ingredients, and environmental samples.

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
79158BC79147BC79145BC
compatibility

for use with GDS PPMX Microsoft Surface Laptop

compatibility

for use with GDS PPMX Microsoft Surface Laptop

compatibility

for use with GDS PPMX Microsoft Surface Laptop

compatibility

BioControl GDS

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

-

packaging

-

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems

manufacturer/tradename

Assurance GDS® PickPen, BioControl Systems 79145




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters