Skip to Content
Merck
All Photos(1)

Key Documents

7910-OP

Millipore

Sodium dodecyl sulfate

OmniPur®

Synonym(s):

OmniPur® Sodium Dodecyl Sulfate, SDS, Sodium Lauryl Sulfate, n-Dodecyl Sulfate, Sodium

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
Inquire
5 KG
Inquire


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
Inquire
5 KG
Inquire

About This Item

Linear Formula:
CH3(CH2)11SO4Na
CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.28


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[S](=O)(=O)([O-])OCCCCCCCCCCCC

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

General description

Suitable for a wide range of molecular biology applications. Molecular biology grade.

Specifications
DNAse: none detected
RNAse: none detected
Protease: none detected
Phosphatase: none detected
Chloride (Cl): ≤0.03%
Copper (Cu): ≤0.0005%
Iron (Fe): ≤0.0001%
Lead (Pb): ≤0.0005%
A260 (0.1 M, water): ≤0.1 AU
A280 (0.1 M, water): ≤0.1 AU
Acidity or alkalinity: Passes test
Purity: ≥99% by assay
Assay (C-12 content): ≥98%
Loss on drying: ≤1.0%
Phosphate (PO4): ≤0.001%
Solubility in alcohol: Passes test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Refining the rheological characteristics of high drug loading ointment via SDS and machine learning.: This research employs sodium dodecyl sulfate (SDS) to optimize the textural properties of pharmaceutical ointments, illustrating the integration of SDS with machine learning techniques for quality control in drug formulation (Liu et al., 2024).

  • Enzymatic and Algebraic Methodology to Determine the Contents of Kunitz and Bowman-Birk Inhibitors and Their Contributions to Total Trypsin or Chymotrypsin Inhibition in Soybeans.: Details a novel use of SDS in enzymatic assays to analyze protein inhibitors in soybeans, offering a methodological advance in agricultural biochemistry (Liu K., 2024).

  • Electrophoresis.: Highlights the pivotal role of SDS in electrophoresis techniques, essential for the separation and analysis of proteins and nucleic acids in biochemical research (Sonagra and Dholariya, 2024).

  • Proteomics reveals the mechanism of protein degradation and its relationship to sensorial and texture characteristics in dry-cured squid during processing.: Utilizes SDS in proteomic studies to explore protein interactions in food processing, linking molecular changes to sensory and textural attributes of seafood (Chen et al., 2024).

Warning

Toxicity: Flammable (J)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service