Skip to Content
Merck
All Photos(2)

Key Documents

79107BC

Millipore

GDS PPMX 4-Column Reagent Reservoir

BioControl, For use with GDS PickPen® PIPETMAX®

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€349.00

€349.00


Estimated to ship on30 May 2025


Request more information

Select a Size

Change View
1 EACH
€349.00

About This Item

UNSPSC Code:
41121522

€349.00


Estimated to ship on30 May 2025


Request more information

description

For use with GDS PICKPEN™ PIPETMAX™ System

manufacturer/tradename

BioControl Systems 79107

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl GDS

storage temp.

room temp

Components

One box, 25 reservoirs/box

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service