Skip to Content
Merck

Skip To

79101BC

GDS PPMX Removable Tray

BioControl Systems, for use with GDS PickPen® PIPETMAX System

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 3.080,00

About This Item

UNSPSC Code:
41121521
NACRES:
NC.81
Manufacturer/tradename:
BioControl Systems

€ 3.080,00


Check Cart for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS PickPen® PIPETMAX System

storage temp.

room temp

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
79105BC79185BC79180BC
manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

compatibility

for use with GDS PickPen® PIPETMAX System

compatibility

for use with GDS PickPen® PIPETMAX System

compatibility

for use with GDS PickPen PIPETMAX System

compatibility

for use with GDS PickPen PIPETMAX System




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters