Skip to Content
Merck
All Photos(1)

Key Documents

73090BC

Millipore

GDS PickPen, 1-Magnet

BioControl®, For use with GDS

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€472.00

€472.00


Estimated to ship on28 May 2025


Request more information

Select a Size

Change View
1 EACH
€472.00

About This Item

UNSPSC Code:
41115866
NACRES:
NA.85

€472.00


Estimated to ship on28 May 2025


Request more information

storage condition

room temperature

Quality Level

application(s)

food and beverages
pathogen testing

compatibility

BioControl®, For use with GDS

Components

One GDS PickPen®

Testing Method

Immunomagnetic separation (IMS)

Legal Information

BioControl is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service