Skip to Content
Merck
All Photos(1)

Key Documents

71284-M

Sigma-Aldrich

pTK-neo DNA - Novagen

For selection of stably transfected mammalian cell lines

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€399.00

€399.00


Estimated to ship on30 May 2025



Select a Size

Change View
20 μG
€399.00

About This Item

UNSPSC Code:
41106508
NACRES:
NA.51

€399.00


Estimated to ship on30 May 2025


Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

−20°C

General description

The pTK-neo vector can select stably transformed mammalian cell lines using G418. A minimal thymidine kinase (TK) promoter controls expression of the neomycin resistance gene. This promoter facilitates selection for stable integration of both the selection plasmid and a cotransfected expression plasmid into transcriptionally active sites in the genome. Use pTK-neo DNA as a cotransfection plasmid with pTandem-1 Mammalian Dual Expression Vector, pTriEx vectors or any mammalian expression vector.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service