Skip to Content
Merck
All Photos(1)

Key Documents

71148-M

Sigma-Aldrich

pETcoco-2 DNA - Novagen

Novagen′s pETcoco-2 expression vector is designed to allow "on command" amplification of vector copy number from single copy (1-2 plasmid copies per cell) to medium copy (20-50 per cell).

Synonym(s):

pETcoco-2 Vector, pETcoco-2 expression vector

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€522.00

€522.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€522.00

About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

€522.00


Estimated to ship on28 May 2025


Request a Bulk Order

conjugate

S-Tag

Quality Level

tag

Herpes Simplex Virus (HSV) tagged
His tagged

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

dry ice
wet ice

storage temp.

−70°C

General description

The pETcoco-2 expression vector is designed to allow "on command" amplification of vector copy number from single copy (1–2 plasmid copies per cell) to medium copy (20–50 per cell). The vector is maintained in the single copy state by utiliziing the oriS and repE elements of the F episome together with the parABC partition determinants. The medium copy state is obtained by employing the RK-2 derived trfA replicator acting at oriV. The switch from single copy to medium copy status is achieved by inducing the expression of the trfA gene with arabinose. The dual replicon nature of these vectors promotes enhanced stability of recombinant plasmids and extremely low basal expression levels in the single copy state, while allowing robust target gene expression typical of the pET vectors. pETcoco-2 contains the ampicillin resistance gene. The sequence is numbered relative to the start of T7 transcription at +1.

The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock). Please contact technical service if you need additional information.

Warning

Toxicity: Standard Handling

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the ′EMD Product′) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: [email protected].

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service