Skip to Content
Merck
All Photos(1)

Key Documents

71097

Sigma-Aldrich

GST•Tag Monoclonal Antibody

Novagen®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€322.00
250 μG
€1,060.00

€322.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€322.00
250 μG
€1,060.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€322.00


Estimated to ship on25 May 2025


Request a Bulk Order

antibody form

purified immunoglobulin

Quality Level

antibody product type

primary antibodies

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description















Sensitive, specific detection of GST•Tag fusion proteins
The GST•Tag Monoclonal Antibody is a mouse monoclonal antibody (IgG1) with high affinity for the 26-kDa glutathione-S-transferase (GST) domain from Schistosoma japonicum. This highly purified antibody is used for detection of fusion proteins containing the GST•Tag expressed with the pET-41 or pET-42 vector series or other GST-encoding vectors. The 50-µg package size provides enough purified antibody to perform 50 Western blots (10 cm × 10 cm).glutathione-S-transferase (GST) domain from Schistosoma japonicum. This highly purified antibody is used for detection of fusion proteins containing the GST•Tag expressed with the pET-41 or pET-42 vector series or other GST-encoding vectors.

The 50-µg package size provides enoughpurified antibody to perform 50 Western blots (10 cm × 10 cm).

Warning

Toxicity: Standard Handling (A)

Other Notes

Fikrig, E. et al. 1990. Science250, 553.
Beekman, J.M., et al. 1994. Gene146, 285.
Poon, R.Y. and Hunt, T. 1994. Anal. Biochem.218, 26.
Toye, B., et al. 1990. Infect. Immunol.58, 3909.
Smith, D.B. and Johnson, K.S. 1988. Gene67, 31.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guido Durian et al.
The Plant cell, 32(5), 1610-1625 (2020-03-01)
Calcium-regulated protein kinases are key components of intracellular signaling in plants that mediate rapid stress-induced responses to changes in the environment. To identify in vivo phosphorylation substrates of CALCIUM-DEPENDENT PROTEIN KINASE1 (CPK1), we analyzed the conditional expression of constitutively active
Keerthika Lohanadan et al.
Cells, 13(1) (2024-01-11)
Synaptopodin-2 (SYNPO2) is a protein associated with the Z-disc in striated muscle cells. It interacts with α-actinin and filamin C, playing a role in Z-disc maintenance under stress by chaperone-assisted selective autophagy (CASA). In smooth muscle cells, SYNPO2 is a
Mengshi Liu et al.
Plants (Basel, Switzerland), 11(11) (2022-06-11)
Histone modifications, such as methylation and demethylation, have crucial roles in regulating chromatin structure and gene expression. Lysine-specific histone demethylases (LSDs) belong to the amine oxidase family, which is an important family of histone lysine demethylases (KDMs), and functions in
Mara E Pitulescu et al.
BMC biochemistry, 10, 16-16 (2009-06-12)
The re-replication inhibitor Geminin binds to several transcription factors including homeodomain proteins, and to members of the polycomb and the SWI/SNF complexes. Here we describe the TATA-binding protein-like factor-interacting protein (TIPT) isoform 2, as a strong binding partner of Geminin.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service