Skip to Content
Merck
All Photos(1)

Key Documents

70548

Sigma-Aldrich

T7Select® 10-3b DNA

Synonym(s):

10-3b DNA, DNA for T7Select, T7Select DNA

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€475.00

€475.00


Estimated to ship on28 May 2025



Select a Size

Change View
10 μG
€475.00

About This Item

UNSPSC Code:
41106616
NACRES:
NA.51

€475.00


Estimated to ship on28 May 2025


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

2-8°C

General description

The T7Select10-3b vector is a protein display vector based on bacteriophage T7. The vector displays an average of 5–15 copies of peptides and proteins up to 1200 aa in size on the surface of the T7 capsid. Target sequences are fused to the C-terminus of the 10B capsid protein near amino acid 348. T7Select10 phage are grown on a complementing host (BLT5403) that supplies large amounts of the 10A capsid protein from a plasmid. Capsids thus contain mostly 10A protein, along with 5–15 copies of 10B fusion protein per virion. The linear genome of T7Select10-3b is 36,249 bp in size. Unique restriction sites are shown on the vector map (TB243).


The T7Select System is sold for research use only. Any commercial use of this Product, including drug discovery or development of commercial products, requires a commercial license from EMD Millipore Corporation. Commercial use shall include but not be limited to (1) use of the Product or its components in manufacturing; (2) use of the Product or its components to provide a service, information, or data to others in exchange for consideration; (3) use of the Product or its components (or any derivatives thereof) for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the Product or its components, whether or not such Product or its components are resold for use in research. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of this Product.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
T7SELECT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service